Learn More
BD Difco™ Dehydrated Culture Media: Noble Agar
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
Brand: BD 214220
Additional Details : CAS Number : 9002-18-0 Weight : 0.11200kg
Description
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
- Solidifying agent
- Free of impurities
Applications:
- Used for electrophoretic and nutritional procedures
Specifications
Noble Agar | |
1.5% | |
1825 Days | |
Dehydrated | |
Bottle |
<30°C | |
Preparing microbiological culture media, for Electrophoretic and nutritional procedures | |
100g | |
ISO/DIS, 11014 |