Learn More
BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems
Product Code. 11797194
Change view
Click to view available options
Type:
EZ Anaerobe Pouch System
EZ CO2 Pouch System
EZ CampyPouch System
Unit Size:
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11797194 EZ CampyPouch System Pack of 20
11777194 EZ Anaerobe Pouch System Pack of 20
11787194 EZ CO2 Pouch System Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11797194 Supplier BD Supplier No. 260685
 Restricted Item
Beräknad leverans: 15-12-2026
Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies provide catalyst-free convenience.
  • One-step resealable pouches
  • The pouches are specially designed to maximize the preservation of the desired environment throughout incubation
  • This heat-free technology minimizes condensate, allowing excellent bacterial colony visualization though the pouch
TRUSTED_SUSTAINABILITY
Specifications
Type EZ CampyPouch System
Closure Type Ziplock
Format Resealable Pouches
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
For Use With (Equipment) BD GasPak EZ Small Incubation Container
Includes Resealable pouches
Product Line GasPak
Quantity 20 Sachets