missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Rat Interleukin-10 (IL-10)
For inhibition of proinflammatory T cell mediated immunity
Brand: BD 555113
Additional Details : Weight : 0.01000kg
Description
- Molecular weight: 18.5kD protein
- Inhibits macrophage-mediated cytokine synthesis
- Suppresses delayed type hypersensitivity response
- Stimulate Th2 cell response
- Helps in elevated antibody production
- Storage buffer: frozen aqueous buffered solution containing BSA
ELISA standard, intracellular block/flow cytometry
Specifications
100μg/mL | |
Frozen aqueous Buffered solution containing BSA | |
Rat IL-10 Recombinant Protein | |
-80°C | |
Recombinant |
ELISA Standard, Intracellular Block/Flow Cytometry | |
18.5 | |
5μg | |
Rat | |
Solution |
For research use only.