Läs mer
BDE 183, TRC
Produktkod. 30134027
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134027 25 mg
30135020 100 mg
17911955 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134027 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB11922525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 207122-16-5
Molekylformel C12 H3 Br7 O
Molekylvikt (g/mol) 722.48
Synonym Benzene, 1,2,3,5-tetrabromo-4-(2,4,5-tribromophenoxy)-, 1,2,3,5-Tetrabromo-4-(2,4,5-tribromophenoxy)benzene, 2,2',3,4,4',5',6-Heptabromodiphenyl ether, BDE 183, PBDE 183, PBDE No. 183
IUPAC-namn 1,2,3,5-tetrabromo-4-(2,4,5-tribromophenoxy)benzene
LEDER Brc1cc(Br)c(Oc2c(Br)cc(Br)c(Br)c2Br)cc1Br
Specifikationer
Kemiskt namn eller material PBDE 183 (2,2',3,4,4',5',6-Heptabromodiphenyl Ether)
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C12H3Br7O/c13-4-1-6(15)9(3-5(4)14)20-12-8(17)2-7(16)10(18)11(12)19/h1-3H
Formel vikt 715.4468
Procent renhet >95%