Läs mer
BDE 28, TRC
Produktkod. 17952055
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17952055 2.5 mg 2.5mg
17949339 25 mg -
17927139 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17952055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1192102.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41318-75-6
Molekylformel C12 H7 Br3 O
Molekylvikt (g/mol) 406.9
Synonym Benzene, 2,4-dibromo-1-(4-bromophenoxy)-, Ether, p-bromophenyl 2,4-dibromophenyl (6CI), 2,4-Dibromo-1-(4-bromophenoxy)benzene, 2,4,4'-Tribromodiphenyl ether, BDE 28, PBDE 28
IUPAC-namn 2,4-dibromo-1-(4-bromophenoxy)benzene
LEDER Brc1ccc(Oc2ccc(Br)cc2Br)cc1
Specifikationer
Kemiskt namn eller material PBDE 28 (2,4,4'-Tribromodiphenyl Ether)
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C12H7Br3O/c13-8-1-4-10(5-2-8)16-12-6-3-9(14)7-11(12)15/h1-7H
Formel vikt 403.8047
Procent renhet >95%