Läs mer
Beclamide 1.0 mg/ml in Acetonitrile, LoGiCal
Produktkod. 30464147
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30464147 1 ml
1 options
Produktkod. 30464147 Leverantör LoGiCal Leverantörsnummer LGCAMP1825.0011
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclamide
CAS 501-68-8
Analyt- eller komponentnamn Beclamide
Frakt skick Ice pack (-20°C)
Molekylformel C10 H12 Cl N O
Certifikat och efterlevnad ISO 17034
Synonym Propionamide, N-benzyl-3-chloro- (6CI,7CI,8CI), Beclamid, Beclamide, Beklamid, Chloracon, Chlorakon, Chloroethylphenamide, Hibicon, N-(3-Chloropropionyl)benzylamine, N-Benzyl-3-chloropropionamide, NSC 67062, Neuracen, Nidrane, Nydran, Nydrane, Posedrine, Seclar, Propanamide, 3-chloro-N-(phenylmethyl)-, N-Benzyl-beta-chloropropanamide, N-Benzyl-beta-chloropropionamide
InChI formel InChI=1S/C10H12ClNO/c11-7-6-10(13)12-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,13)
Lösningstyp Single Solution
LEDER ClCCC(=O)NCc1ccccc1
IUPAC-namn N-benzyl-3-chloropropanamide
Lösningsmedel eller matris Acetonitrile
Industry Type Clinical Testing
Molekylvikt (g/mol) 197.66
Kvantitet 1 ml
Formel vikt 197.0607 g/mol