Läs mer
Beclamide, LoGiCal
Produktkod. 30457713
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30457713 10 mg
1 options
Produktkod. 30457713 Leverantör LoGiCal Leverantörsnummer LGCFOR1825.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclamide
CAS 501-68-8
Analyt- eller komponentnamn Beclamide
Frakt skick Room Temperature
Molekylformel C10 H12 Cl N O
Certifikat och efterlevnad ISO 9001
Synonym Propionamide, N-benzyl-3-chloro- (6CI,7CI,8CI), Beclamid, Beclamide, Beklamid, Chloracon, Chlorakon, Chloroethylphenamide, Hibicon, N-(3-Chloropropionyl)benzylamine, N-Benzyl-3-chloropropionamide, NSC 67062, Neuracen, Nidrane, Nydran, Nydrane, Posedrine, Seclar, Propanamide, 3-chloro-N-(phenylmethyl)-, N-Benzyl-beta-chloropropanamide, N-Benzyl-beta-chloropropionamide
InChI formel InChI=1S/C10H12ClNO/c11-7-6-10(13)12-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,13)
Lösningstyp Neat
LEDER ClCCC(=O)NCc1ccccc1
IUPAC-namn N-benzyl-3-chloropropanamide
Industry Type Clinical Testing
Molekylvikt (g/mol) 197.66
Kvantitet 10 mg
Formel vikt 197.0607 g/mol