Läs mer
Beclamide, Mikromol™
Produktkod. 30051532
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30051532 100 mg
1 options
Produktkod. 30051532 Leverantör Mikromol Leverantörsnummer MM1825.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Beclamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclamide
CAS 501-68-8
Analyt- eller komponentnamn Beclamide
Frakt skick Room Temperature
Molekylformel C10 H12 Cl N O
Certifikat och efterlevnad ISO 17025
Synonym Propionamide, N-benzyl-3-chloro- (6CI,7CI,8CI), Beclamid, Beclamide, Beklamid, Chloracon, Chlorakon, Chloroethylphenamide, Hibicon, N-(3-Chloropropionyl)benzylamine, N-Benzyl-3-chloropropionamide, NSC 67062, Neuracen, Nidrane, Nydran, Nydrane, Posedrine, Seclar, Propanamide, 3-chloro-N-(phenylmethyl)-, N-Benzyl-beta-chloropropanamide, N-Benzyl-beta-chloropropionamide
InChI formel InChI=1S/C10H12ClNO/c11-7-6-10(13)12-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,13)
LEDER ClCCC(=O)NCc1ccccc1
IUPAC-namn N-benzyl-3-chloropropanamide
Molekylvikt (g/mol) 197.66
Kvantitet 100 mg
Formel vikt 197.0607 g/mol