Läs mer
Beclamide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Beclamide |
| CAS | 501-68-8 |
| Molekylformel | C10 H12 Cl N O |
| Kvantitet | 25 g |
| Synonym | Propionamide, N-benzyl-3-chloro- (6CI,7CI,8CI), Beclamid, Beclamide, Beklamid, Chloracon, Chlorakon, Chloroethylphenamide, Hibicon, N-(3-Chloropropionyl)benzylamine, N-Benzyl-3-chloropropionamide, N-Benzyl-β-chloropropanamide, N-Benzyl-β-chloropropionamide, NSC 67062, Neuracen, Nidrane, Nydran, Nydrane, Posedrine, Seclar, Propanamide, 3-chloro-N-(phenylmethyl)- |
| InChI formel | InChI=1S/C10H12ClNO/c11-7-6-10(13)12-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,13) |
| LEDER | ClCCC(=O)NCc1ccccc1 |
| IUPAC-namn | N-benzyl-3-chloropropanamide |
| Molekylvikt (g/mol) | 197.66 |
| Formel vikt | 197.0607 |