Läs mer
Beclometasone 17-Propionate, LoGiCal
Produktkod. 30460217
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30460217 10 mg
1 options
Produktkod. 30460217 Leverantör LoGiCal Leverantörsnummer LGCFOR0199.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclometasone 17-Propionate
CAS 5534-18-9
Analyt- eller komponentnamn Beclometasone 17-Propionate
Frakt skick Room Temperature
Molekylformel C25 H33 Cl O6
Certifikat och efterlevnad ISO 9001
Synonym 9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate, Beclometasone Dipropionate Imp. H (EP), Beclometasone 17-Propionate, Beclomethasone 17-monopropionate, Beclomethasone 17-propionate, (11beta,16beta)-9-Chloro-11,21-dihydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Chloro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 17-propionate, CCI 2382, Beclometasone Dipropionate Related Compound H, Beclometasone Dipropionate Impurity H
InChI formel InChI=1S/C25H33ClO6/c1-5-21(31)32-25(20(30)13-27)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,26)19(29)12-23(18,25)4/h8-9,11,14,17-19,27,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
Lösningstyp Neat
LEDER CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C)C(=O)CO
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
Industry Type Clinical Testing
Molekylvikt (g/mol) 464.98
Kvantitet 10 mg
Formel vikt 464.1966 g/mol