Läs mer
Beclomethasone 11,17,21-Tripropionate, TRC
Produktkod. 30333350
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30333350 1 g
1 options
Produktkod. 30333350 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1310751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclomethasone 11,17,21-Tripropionate
CAS 1709825-83-1
Molekylformel C31 H41 Cl O8
Kvantitet 1 g
Synonym (8S,9R,10S,11S,13S,14S,16S,17R)-9-Chloro-10,13,16-trimethyl-3-oxo-17-(2-(propionyloxy)acetyl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-11,17-diyl Dipropionate, Beclomethasone Dipropionate EP Impurity S, 9-Chloro-16β-methyl-3,20-dioxo-pregna-1,4-diene-11β,17,21-triyl tripropanoate, Beclometasone tripropionate, Beclometasone Tripropionate
InChI formel InChI=1S/C31H41ClO8/c1-7-25(35)38-17-23(34)31(40-27(37)9-3)18(4)14-22-21-11-10-19-15-20(33)12-13-28(19,5)30(21,32)24(16-29(22,31)6)39-26(36)8-2/h12-13,15,18,21-22,24H,7-11,14,16-17H2,1-6H3/t18-,21-,22-,24-,28-,29-,30-,31-/m0/s1
LEDER O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2(Cl)[C@@H](OC(CC)=O)C[C@@]4(C)[C@@]3([H])C[C@H](C)[C@]4(OC(CC)=O)C(COC(CC)=O)=O)=C1
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-10,13,16-trimethyl-3-oxo-11,17-di(propanoyloxy)-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molekylvikt (g/mol) 577.11
Formel vikt 576.249