Läs mer
Beclomethasone Hemisuccinate, TRC
Produktkod. 30342225
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30342225 10 mg
1 options
Produktkod. 30342225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB13104010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclomethasone Hemisuccinate
CAS 75899-57-9
Molekylformel C26H33ClO8
Kvantitet 10 mg
Synonym (11beta,16beta)-21-(3-Carboxy-1-oxopropoxy)-9-chloro-11,17-dihydroxy-16-methyl-pregna-1,4-diene-3,20-dione
InChI formel InChI=1S/C26H33ClO8/c1-14-10-18-17-5-4-15-11-16(28)8-9-23(15,2)25(17,27)19(29)12-24(18,3)26(14,34)20(30)13-35-22(33)7-6-21(31)32/h8-9,11,14,17-19,29,34H,4-7,10,12-13H2,1-3H3,(H,31,32)/t14-,17-,18-,19-,23-,24-,25-,26-/m0/s1
LEDER C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)CCC(=O)O
IUPAC-namn 4-[2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid
Molekylvikt (g/mol) 508.99
Formel vikt 508.1864