Läs mer
Beclotiamine Hydrochloride, TRC
Produktkod. 30012856
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012856 2.5 g
17951045 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1195002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7275-24-3
Molekylformel C12 H16 Cl N4 S . Cl . Cl H
Molekylvikt (g/mol) 355.71
Synonym Thiamine Hydrochloride Imp. C (EP), Thiamine Imp. C (EP), 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-chloroethyl)-4-methylthiazolium Chloride Hydrochloride, Thiamine Hydrochloride Imp. C (EP) as Chloride Hydrochloride, Chlorothiamine Chloride Hydrochloride, Thiamine Hydrochloride Impurity C as Chloride Hydrochloride, Thiamine Impurity C as Chloride Hydrochloride
IUPAC-namn 5-[[5-(2-chloroethyl)-4-methyl-1,3-thiazol-3-ium-3-yl]methyl]-2-methylpyrimidin-4-amine;chloride;hydrochloride
LEDER Cl.[Cl-].Cc1ncc(C[n+]2csc(CCCl)c2C)c(N)n1
Specifikationer
Kemiskt namn eller material 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-chloroethyl)-4-methylthiazolium Chloride Hydrochloride (Chlorothiamine Chloride Hydrochloride)
Analyt- eller komponentnamn 3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-chloroethyl)-4-methylthiazolium Chloride Hydrochloride (Chlorothiamine Chloride Hydrochloride)
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C12H16ClN4S.2ClH/c1-8-11(3-4-13)18-7-17(8)6-10-5-15-9(2)16-12(10)14;;/h5,7H,3-4,6H2,1-2H3,(H2,14,15,16);2*1H/q+1;;/p-1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 2.5 g
Formel vikt 354.024
Procent renhet >95%