Läs mer
Beeswax, TRC
Produktkod. 30347726
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30347726 5 g
1 options
Produktkod. 30347726 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1194505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beeswax
CAS 8012-89-3
Kvantitet 5 g
Synonym White Beeswax, AR 100, Aquacer 560, BWS 30, Bee, wax of, Beeswax CO 100
LEDER CC(O[C@H]1C(OC(C)=O)O[C@@H](C1)COC(C2=CC=CC=C2)=O)=O