Läs mer
Behenic Acid Ethyl Ester, TRC
Produktkod. 30342435
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30342435 1 g
1 options
Produktkod. 30342435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1195851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Docosanoic acid ethyl ester
CAS 5908-87-2
Molekylformel C24 H48 O2
Kvantitet 1 g
Synonym Docosanoic Acid Ethyl Ester, Behenic acid ethyl ester, Ethyl behenate, Ethyl docosanoate
InChI formel InChI=1S/C24H48O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26-4-2/h3-23H2,1-2H3
LEDER CCCCCCCCCCCCCCCCCCCCCC(=O)OCC
IUPAC-namn ethyl docosanoate
Molekylvikt (g/mol) 368.64
Formel vikt 368.3654