Läs mer
Behentrimonium Chloride (>80%), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Docosyltrimethylammonium Chloride |
| CAS | 17301-53-0 |
| Molekylformel | C25 H54 N . Cl |
| Kvantitet | 50 mg |
| Synonym | ,N,N-Trimethyldocosan-1-aminium Chloride, Arquad 22-80, Behenyltrimethylammonium Chloride, Catinal DC 80, Cation VB 800, Docosyltrimethylammonium chloride, Econol TM 22, Gemanin KDM-P, Genamin DDMP, Genamin GDMP, Incroquat Behenyl TMC, Incroquat Behenyl TMC 85, Incroquat TMC 95, Kemamine Q 2803C, N-Docosyl-N,N,N-trimethylammonium chloride, Neoscoap CN 30SF, Nissan Cation VB, Quartamin 2285E-E, Trimethylbehenylammonium chloride, Varisoft BT 85 |
| InChI formel | InChI=1S/C25H54N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(2,3)4;/h5-25H2,1-4H3;1H/q+1;/p-1 |
| LEDER | [Cl-].CCCCCCCCCCCCCCCCCCCCCC[N+](C)(C)C |
| IUPAC-namn | docosyl(trimethyl)azanium;chloride |
| Molekylvikt (g/mol) | 404.156 |
| Formel vikt | 403.394 |