missing translation for 'onlineSavingsMsg'
Läs mer

Behentrimonium Chloride (>80%), TRC

Produktkod. 30347792
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30347792 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30347792 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB100100250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Docosyltrimethylammonium Chloride
CAS 17301-53-0
Molekylformel C25 H54 N . Cl
Kvantitet 250 mg
Synonym ,N,N-Trimethyldocosan-1-aminium Chloride, Arquad 22-80, Behenyltrimethylammonium Chloride, Catinal DC 80, Cation VB 800, Docosyltrimethylammonium chloride, Econol TM 22, Gemanin KDM-P, Genamin DDMP, Genamin GDMP, Incroquat Behenyl TMC, Incroquat Behenyl TMC 85, Incroquat TMC 95, Kemamine Q 2803C, N-Docosyl-N,N,N-trimethylammonium chloride, Neoscoap CN 30SF, Nissan Cation VB, Quartamin 2285E-E, Trimethylbehenylammonium chloride, Varisoft BT 85
InChI formel InChI=1S/C25H54N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(2,3)4;/h5-25H2,1-4H3;1H/q+1;/p-1
LEDER [Cl-].CCCCCCCCCCCCCCCCCCCCCC[N+](C)(C)C
IUPAC-namn docosyl(trimethyl)azanium;chloride
Molekylvikt (g/mol) 404.156
Formel vikt 403.394
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.