Läs mer
Behenyl Behenate, TRC
Produktkod. 30331001
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30331001 100 mg
1 options
Produktkod. 30331001 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB131153100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Behenyl Behenate
CAS 17671-27-1
Molekylformel C44 H88 O2
Kvantitet 100 mg
Synonym Behenyl Behenate, Behenic acid behenyl ester, Behenic behenate, Behenyl behenate, Docosyl behenate, Electol WEP 3, Esprix N 252, Exceparl BB, Kester Wax 72, Nissan Electol WEP 3, Nissan Unister M 2222SL, Pelemol BB, Rikaflow BB 62, SP-C 44 Ester Wax, Unister M 2222SL, WEP 3, n-Docosyl n-docosanoate
InChI formel InChI=1S/C44H88O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-43-46-44(45)42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-43H2,1-2H3
LEDER O=C(OCCCCCCCCCCCCCCCCCCCCCC)CCCCCCCCCCCCCCCCCCCCC
IUPAC-namn docosyl docosanoate
Molekylvikt (g/mol) 649.168
Formel vikt 648.678