Learn More
SP Bel-Art™ Bel-Art Products Sterileware Pharma Scoops
Ideal for both production work and laboratory use, Pharma Scoops are used by pharmaceutical, food, cosmetic, and chemical companies
Brand: SP Bel-Art™ F36910-0125
Additional Details : Weight : 485.00000kg
Description
- Bright red color reduces chance of scoop being left behind in a sample; can serve as code to avoid cross contamination; can provide contrasting background for light colored samples
- High handle location keeps hand further from sample
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
125 mL | |
100/Pk. | |
10.4 cm |
Red | |
9.2 x 6.1 x 4.5 cm |