Learn More
Bel-Art™ SP Scienceware™ DNA Band Excisors
For excising DNA bands or purified plasmid DNA preps from agarose/polyacrylamide gels
Brand: Bel-Art™ F36970-0010
Additional Details : Weight : 0.11700kg
Description
- Double-ended polystyrene tool with thin blade and spatula
- Used in laboratories and educational settings
- Reduces exposure of nucleic acids to UV light and prevents scratches to transilluminator screens
- Discarded in standard trash receptacles
Specifications
Laboratories and educational settings | |
Dna Band Excisor |