Learn More
Bel-Art™ SP Scienceware™ Glove Box Economy Sleeve Gloves
Catalog Number 11757385
Click to view available options
Size:
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy
One piece, bonded neoprene protection against liquid or gas penetration
  • Single piece glove with sleeve
  • For glove boxes with 8 in. glove ports
  • Requires clamping rings (19-821-983) to hold gloves in place
  • Sold in pairs