Learn More
Bel-Art™ SP Scienceware™ Micro-Mill™ Grinder Blades
Replacement blades for MicroMill grinder
Brand: Bel-Art™ H37257-0000
Additional Details : Weight : 0.01000kg
Description
Stainless-steel or hard-faced blades.
Certifications
CE Marked
Specifications
Replacement Blade | |
MicroMill grinder | |
12,000 rpm |
Ideal for processing minerals, pharmaceuticals, pigment blends and other solid samples | |
Stainless Steel |