Learn More
Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars in Colors
Registered ISO 9001:2008. SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars in Colors are the most commonly used shape.
2375.00 SEK
Product Code | Brand | Color | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
11752685
|
Bel-Art™
F37109-0034 |
Red | 3.2 mm | 12.7 mm |
2375.00 SEK
Pack of 50 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||
Description
- PTFE-coated
- With pivot ring
- Assorted sizes and colors
Specifications
Octagon |