missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Poxygrid™ Wire Test Tube Racks: 18-20mm

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

Brand:  Bel-Art™ F18762-0002

Additional Details : Weight : 0.24760kg

 View more versions of this product

Product Code. 15510032

  • 428.00 SEK / Each
In Stock
to see stock.
Add to basket

You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.

Description

Description

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable

Specifications

Specifications

Test Tube Rack
Yellow
Epoxy Coated Steel
9.6 x 4.25 x 3.25 in. (243 x 110 x 83mm)
9.6 in.
4.25 in.
3.25 in.
Autoclavable
Tubes (16 to 20 mm)
40
243 mm
110 mm
83 mm
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers