Läs mer
Bel Art™ Secador™ 1.0 Bärväska exsickator
Handla allt SP Scienceware Produkter
Förpackningsstorlek:
Varje
Färg:
Blått
Beskrivning
- Durastar™ co-polyester construction blocks 99% of UV light and is resistant to staining, crazing and chemical attack
- Large door maximizes access to interior space and has a patented seal design, tab latches and security loops for a padlock or tamper-evident seals; opens top to bottom
- Includes built-in suitcase style carrying handle and two removable shelves
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view dial hygrometer located on pull down oven style door
- Can be used with desiccant cartridges F42048-0065 or F42046-0000
- 16.3 D x 8.4 W x 14.7 in. H (41.4 x 21.3 x 37.3 cm)
- Internal volume: 0.75 cu. ft.
- U.S. Patents: 6,834,920; 7,318,630 and D474,626
Specifikationer
Specifikationer
| Skåp stil | Front-loading |
| Material | Copolymer |
| Produkttyp | Carrying Case Desiccator |
| För användning med (applikation) | For dry storage of moisture and UV sensitive products |
| Volym (engelska) | 0.75 cu. ft |
| Höjd (engelska) | 14.7 in. |
| Höjd (metrisk) | 37.3 cm |