missing translation for 'onlineSavingsMsg'
Läs mer

Belotecan Hydrochloride, TRC

Produktkod. 30347607
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30347607 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30347607 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB131300100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Belotecan Hydrochloride
CAS 213819-48-8
Molekylformel C25 H27 N3 O4 . Cl H
Kvantitet 100 mg
Synonym 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4-hydroxy-11-[2-[(1-methylethyl)amino]ethyl]-, hydrochloride (1:1), (4S)-, 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4-hydroxy-11-[2-[(1-methylethyl)amino]ethyl]-, monohydrochloride, (4S)- (9CI), (S)-4-Ethyl-4-hydroxy-11-(2-(isopropylamino)ethyl)-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride, Belotecan hydrochloride, CKD 602
InChI formel InChI=1S/C25H27N3O4.ClH/c1-4-25(31)19-11-21-22-17(12-28(21)23(29)18(19)13-32-24(25)30)15(9-10-26-14(2)3)16-7-5-6-8-20(16)27-22;/h5-8,11,14,26,31H,4,9-10,12-13H2,1-3H3;1H/t25-;/m0./s1
LEDER Cl.CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4c(CCNC(C)C)c5ccccc5nc34)C2=O
IUPAC-namn (19S)-19-ethyl-19-hydroxy-10-[2-(propan-2-ylamino)ethyl]-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2,4,6,8,10,15(20)-heptaene-14,18-dione;hydrochloride
Molekylvikt (g/mol) 469.96
Formel vikt 469.1768
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.