Läs mer
Belotecan Hydrochloride, TRC
Produktkod. 30329524
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30329524 10 mg
1 options
Produktkod. 30329524 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB13130010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Belotecan Hydrochloride
CAS 213819-48-8
Molekylformel C25 H27 N3 O4 . Cl H
Kvantitet 10 mg
Synonym 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4-hydroxy-11-[2-[(1-methylethyl)amino]ethyl]-, hydrochloride (1:1), (4S)-, 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4-hydroxy-11-[2-[(1-methylethyl)amino]ethyl]-, monohydrochloride, (4S)- (9CI), (S)-4-Ethyl-4-hydroxy-11-(2-(isopropylamino)ethyl)-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride, Belotecan hydrochloride, CKD 602
InChI formel InChI=1S/C25H27N3O4.ClH/c1-4-25(31)19-11-21-22-17(12-28(21)23(29)18(19)13-32-24(25)30)15(9-10-26-14(2)3)16-7-5-6-8-20(16)27-22;/h5-8,11,14,26,31H,4,9-10,12-13H2,1-3H3;1H/t25-;/m0./s1
LEDER Cl.CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4c(CCNC(C)C)c5ccccc5nc34)C2=O
IUPAC-namn (19S)-19-ethyl-19-hydroxy-10-[2-(propan-2-ylamino)ethyl]-17-oxa-3,13-diazapentacyclo[11.8.0.02,11.04,9.015,20]henicosa-1(21),2,4,6,8,10,15(20)-heptaene-14,18-dione;hydrochloride
Molekylvikt (g/mol) 469.96
Formel vikt 469.1768