Läs mer
Bempedoic Acid, TRC
Produktkod. 30136829
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136829 100 mg
17990715 250 mg
17910375 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136829 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB119700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 738606-46-7
Molekylformel C19H36O5
Molekylvikt (g/mol) 344.49
Synonym 8-Hydroxy-2,2,14,14-tetramethylpentadecanedioic Acid, ESP 55016, ETC 1002:
IUPAC-namn 8-hydroxy-2,2,14,14-tetramethylpentadecanedioic acid
LEDER CC(C)(CCCCCC(O)CCCCCC(C)(C)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Bempedoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C19H36O5/c1-18(2,16(21)22)13-9-5-7-11-15(20)12-8-6-10-14-19(3,4)17(23)24/h15,20H,5-14H2,1-4H3,(H,21,22)(H,23,24)
Formel vikt 344.2563
Procent renhet >95%