Läs mer
Benazepril tert-Butyl Ester, TRC
Produktkod. 30336673
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30336673 1 g
1 options
Produktkod. 30336673 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1198501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benazepril tert-Butyl Ester
CAS 109010-61-9
Molekylformel C28H36N2O5
Kvantitet 1 g
Synonym 1H-1-Benzazepine-1-acetic acid, 3-[[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, 1,1-dimethylethyl ester, (3S)-, 1H-1-Benzazepine-1-acetic acid, 3-[[1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, 1,1-dimethylethyl ester, [S-(R*,R*)]-, 1,1-Dimethylethyl (3S)-3-[[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetate, Benazepril tert-Butyl Ester
InChI formel InChI=1S/C28H36N2O5/c1-5-34-27(33)23(17-15-20-11-7-6-8-12-20)29-22-18-16-21-13-9-10-14-24(21)30(26(22)32)19-25(31)35-28(2,3)4/h6-14,22-23,29H,5,15-19H2,1-4H3/t22-,23-/m0/s1
LEDER CCOC(=O)[C@H](CCc1ccccc1)N[C@H]2CCc3ccccc3N(CC(=O)OC(C)(C)C)C2=O
IUPAC-namn ethyl (2S)-2-[[(3S)-1-[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]-2-oxo-4,5-dihydro-3H-1-benzazepin-3-yl]amino]-4-phenylbutanoate
Molekylvikt (g/mol) 480.6
Formel vikt 480.2624