Beskrivning
- Max temp 850°C
- Horizontal air circulation with optimum distribution through stainless steel baffles
- Swing door hinged on the right
Specifikationer
Specifikationer
| Controllertyp | B510 |
| Kapacitet (metrisk) | 15 l |
| Temperatur (metrisk) | 850°C |
| Typ | Furnace |