Learn More
Eppendorf™ Benchtop Microcentrifuge
Choose from a range of models with different rotors for a variety of centrifugation applications. Eppendorf™ Benchtop Microcentrifuges are ideal for safe centrifugation of hazardous samples.
24100.00 SEK
Specifications
Display Digital
Rotor Included Yes
Safety Features Aerosol-tight
View More Specs
Products 1
Product Code Brand Controller Type Height (Metric) Exterior Product Line Price Quantity & Availability  
Product Code Brand Controller Type Height (Metric) Exterior Product Line Price Quantity & Availability  
10216522 View accessory products Eppendorf™
5424000410
Knob 5424 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Standard rotor can accommodate 1.5mL/2.0mL tubes
  • PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor for molecular and biological laboratories
  • Automatic lid opening at the end of the run – prevents sample warming and allows easy access to samples (Centrifuge 5418 and 5424)
The 5418 and 5418 R models are 18 place microcentrifuges for 1.5mL to 2.0mL tubes.
Small footprint
  • Quiet operation without rotor lid
  • Aerosol-tight rotor as standard
  • Converts rpm/RCF
  • Short spin key For research and clinical laboratories
Centrifuge 5424 and 5424 R feature 24–place capacity and speed up to 21,130 × g
  • Available as a rotary knob version for fast parameter selection, or keypad version for easy cleaning
  • OptiBowl design for quiet operation, even without a rotor lid
  • Soft-touch one-finger lid closure for ergonomic operation
Specifications
Specifications
Digital
Yes
Aerosol-tight