missing translation for 'onlineSavingsMsg'
Learn More

BEND5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20931-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16850515

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

BEND5
Polyclonal
Unconjugated
BEND5
BEN domain containing 5, BEND5, C1orf165
Rabbit
Antigen Affinity Chromatography
RUO
67621, 79656
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7L4P6, Q8C6D4
Bend5
BEND5 Fusion Protein Ag14930
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers