Läs mer
Bendamustine D-Mannitol Ester, TRC
Produktkod. 30134940
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134940 50 mg
30007651 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134940 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB13253550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1869075-89-7
Molekylformel C22 H33 Cl2 N3 O7
Molekylvikt (g/mol) 522.42
Synonym Mannitol-1-yl 4-[5-[bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazol-2-yl]butanoate, [(2R,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexyl] 4-[5-[bis(2-chloroethyl)amino]-1-methyl-benzimidazol-2-yl]butanoate
IUPAC-namn [(2R,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl] 4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoate
LEDER Cn1c(CCCC(=O)OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)nc2cc(ccc12)N(CCCl)CCCl
Specifikationer
Kemiskt namn eller material Bendamustine D-Mannitol ester
Analyt- eller komponentnamn Bendamustine D-Mannitol ester
Frakt skick Room Temperature
InChI formel InChI=1S/C22H33Cl2N3O7/c1-26-16-6-5-14(27(9-7-23)10-8-24)11-15(16)25-19(26)3-2-4-20(31)34-13-18(30)22(33)21(32)17(29)12-28/h5-6,11,17-18,21-22,28-30,32-33H,2-4,7-10,12-13H2,1H3/t17-,18-,21-,22-/m1/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 50 mg
Formel vikt 521.1696