Läs mer
Bendamustine Dimer Impurity, TRC
Produktkod. 30333435
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30333435 25 mg
1 options
Produktkod. 30333435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB13252025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bendamustine dimer
CAS 1228551-91-4
Molekylformel C32 H41 Cl3 N6 O4
Kvantitet 25 mg
Synonym 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, 2-[[2-(3-carboxypropyl)-1-methyl-1H-benzimidazol-5-yl](2-chloroethyl)amino]ethyl ester, 4-[5-({2-[(4-{5-[Bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazol-2-yl}butanoyl)oxy]ethyl}(2-chloroethyl)amino)-1-methyl-1H-benzimidazol-2-yl]butanoic acid, 4-[5-[2-[4-[5-[Bis(2-chloroethyl)amino]-1-methyl-benzimidazol-2-yl]butanoyloxy]ethyl-(2-chloroethyl)amino]-1-methyl-benzimidazol-2-yl]butanoic acid, Bendamustine dimer, Bendamustine chloro dimer, Bendamustine Rel. Comp. H (USP)
InChI formel InChI=1S/C32H41Cl3N6O4/c1-38-28-12-10-24(22-26(28)36-29(38)5-3-7-31(42)43)41(18-15-35)19-20-45-32(44)8-4-6-30-37-25-21-23(9-11-27(25)39(30)2)40(16-13-33)17-14-34/h9-12,21-22H,3-8,13-20H2,1-2H3,(H,42,43)
LEDER Cn1c(CCCC(=O)O)nc2cc(ccc12)N(CCCl)CCOC(=O)CCCc3nc4cc(ccc4n3C)N(CCCl)CCCl
IUPAC-namn 4-[5-[2-[4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoyloxy]ethyl-(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid
Molekylvikt (g/mol) 680.06
Formel vikt 678.2255