Läs mer
Bendamustine Isopropyl Ester, TRC
Produktkod. 30332725
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30332725 10 mg
1 options
Produktkod. 30332725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB13251010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bendamustine Isopropyl Ester
CAS 1313020-25-5
Molekylformel C19 H27 Cl2 N3 O2
Kvantitet 10 mg
Synonym 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, 1-methylethyl ester, 1-Methylethyl 5-[bis(2-chloroethyl)amino]-1-methyl-1H-Benzimidazole-2-butanoate, Bendamustine Isopropyl Ester, Bendamustine 1-Methylethyl Ester
InChI formel InChI=1S/C19H27Cl2N3O2/c1-14(2)26-19(25)6-4-5-18-22-16-13-15(7-8-17(16)23(18)3)24(11-9-20)12-10-21/h7-8,13-14H,4-6,9-12H2,1-3H3
LEDER CC(C)OC(=O)CCCc1nc2cc(ccc2n1C)N(CCCl)CCCl
IUPAC-namn propan-2-yl 4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoate
Molekylvikt (g/mol) 400.34
Formel vikt 399.148