Läs mer
Bendazac L-Lysine, MedChemExpress
Produktkod. 16458856
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16458856 100 mg
1 options
Produktkod. 16458856 Leverantör Medchem Express Leverantörsnummer HYB2165
MedChemExpress Bendazac L-Lysine is one of agents that have been introduced for the management of cataracts, protecting the level of vision in patients, thus delaying the need for surgical intervention.
Bendazac L-Lysine is one of agents that have been introduced for the management of cataracts, protecting the level of vision in patients, thus delaying the need for surgical intervention.
Specifikationer
Kemiskt namn eller material Bendazac L-Lysine
CAS 81919-14-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C22H28N4O5
Löslighetsinformation H2O : 25 mg/mL (58.35 mM; Need ultrasonic)
LEDER O=C(O)COC1=NN(CC2=CC=CC=C2)C3=C1C=CC=C3.N[C@@H](CCCCN)C(O)=O
Molekylvikt (g/mol) 428.48
Kvantitet 100 mg
Formel vikt 428.48
Procent renhet 99.82%
Kvalitet Research
Fysisk form Solid