Läs mer
Bendiocarb, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Bendiocarb |
| CAS | 22781-23-3 |
| Molekylformel | C11 H13 N O4 |
| Kvantitet | 500 mg |
| Synonym | 1,3-Benzodioxol-4-ol, 2,2-dimethyl-, methylcarbamate (9CI), Carbamic acid, methyl-, 2,3-(isopropylidenedioxy)phenyl ester (8CI), Phenol, 2,3-(isopropylidenedioxy)-, methylcarbamate (8CI), 2,2-Dimethyl-1,3-benzodioxol-4-yl N-methylcarbamate, 2,2-Dimethyl-1,3-benzodioxol-4-yl methylcarbamate, Bencarbate, Bendiocarb, Dycarb, Ficam, Ficam 80W, Ficam W, Fisons NC 6897, Garvox, Multamat, NC 6897, Niomil, Seedox, Seedox (insecticide), Seedox 80W, Tatoo, Tattoo, Turcam |
| InChI formel | InChI=1S/C11H13NO4/c1-11(2)15-8-6-4-5-7(9(8)16-11)14-10(13)12-3/h4-6H,1-3H3,(H,12,13) |
| LEDER | CNC(=O)Oc1cccc2OC(C)(C)Oc12 |
| IUPAC-namn | (2,2-dimethyl-1,3-benzodioxol-4-yl) N-methylcarbamate |
| Molekylvikt (g/mol) | 223.23 |
| Formel vikt | 223.0845 |