Läs mer
Benfluralin, MedChemExpress
Produktkod. 16404647
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16404647 50 mg
1 options
Produktkod. 16404647 Leverantör Medchem Express Leverantörsnummer HYB2045
MedChemExpress Benfluralin is a kind of herbicide and an agrochemical which can be used as a pre-emergence herbicide used for the control of grass and other weeds in a range of food and non-food crops.
Benfluralin is a kind of herbicide and an agrochemical which can be used as a pre-emergence herbicide used for the control of grass and other weeds in a range of food and non-food crops.
Specifikationer
Kemiskt namn eller material Benfluralin
CAS 1861-40-1
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C13H16F3N3O4
Kvantitet 50 mg
Löslighetsinformation DMSO : 150 mg/mL (447.39 mM; Need ultrasonic)
LEDER FC(C1=CC([N+]([O-])=O)=C(N(CCCC)CC)C([N+]([O-])=O)=C1)(F)F
Molekylvikt (g/mol) 335.28
Formel vikt 335.28
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid