Läs mer
Benfluralin, TRC
Produktkod. 30345689
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30345689 500 mg
30348721 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30345689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB120100500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benfluralin
CAS 1861-40-1
Molekylformel C13 H16 F3 N3 O4
Kvantitet 500 mg
Synonym p-Toluidine, N-butyl-N-ethyl-α,α,α-trifluoro-2,6-dinitro- (7CI,8CI), N-Butyl-N-ethyl-2,6-dinitro-4-(trifluoromethyl)benzenamine, 2,6-Dinitro-4-trifluoromethyl-N-n-butyl-N-ethylaniline, 4-Trifluoromethyl-2,6-dinitro-N-ethyl-N-butylaniline, Balan, Balfin, Banafine, Benafine, Benalan, Benefex, Benefin, Benefine, Benephin, Benfluralin, Benfluraline, Bethrodine, Bhulan, Binnell, Bonalan, Carpidor, EL 110, Emblem, Flubalex, N-Butyl-2,6-dinitro-N-ethyl-4-trifluoromethylaniline, N-Butyl-N-ethyl-2,6-dinitro-4-trifluoromethylaniline, N-Butyl-N-ethyl-4-trifluoromethyl-2,6-trinitroaniline, N-Butyl-N-ethyl-α,α,α-trifluoro-2,6-dinitro-p-toluidine, N-Ethyl-N-butyl-2,6-dinitro-4-trifluoromethylaniline, Pel-Tech, Quilan, α,α,α,Trifluoro-2,6-dinitro-N,N-ethylbutyl-p-toluidine
InChI formel InChI=1S/C13H16F3N3O4/c1-3-5-6-17(4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8H,3-6H2,1-2H3
LEDER CCCCN(CC)c1c(cc(cc1[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-]
IUPAC-namn N-butyl-N-ethyl-2,6-dinitro-4-(trifluoromethyl)aniline
Molekylvikt (g/mol) 335.28
Formel vikt 335.1093