Läs mer
Benmijunzhi, TRC
Produktkod. 30336715
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30336715 50 mg
1 options
Produktkod. 30336715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB85236050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benmijunzhi
CAS 852369-40-5
Molekylformel C20H22O4
Kvantitet 50 mg
Synonym Benmijunzhi, Benzeneacetic acid, 2-[(2,5-dimethylphenoxy)methyl]-α-(methoxymethylene)-, methyl ester, (αE)-, Methyl (αE)-2-[(2,5-dimethylphenoxy)methyl]-α-(methoxymethylene)benzeneacetate, Benmijunzhi, ZJ 0712, Methyl (alphaE)-2-[(2,5-Dimethylphenoxy)methyl]-alpha-(methoxymethylene)benzeneacetate, ZJ 0712
InChI formel InChI=1S/C20H22O4/c1-14-9-10-15(2)19(11-14)24-12-16-7-5-6-8-17(16)18(13-22-3)20(21)23-4/h5-11,13H,12H2,1-4H3/b18-13+
LEDER CO\C=C(\C(=O)OC)/c1ccccc1COc2cc(C)ccc2C
IUPAC-namn methyl (E)-2-[2-[(2,5-dimethylphenoxy)methyl]phenyl]-3-methoxyprop-2-enoate
Molekylvikt (g/mol) 326.39
Formel vikt 326.1518