Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benomyl |
| Förpackning | Glass Bottle |
| CAS | 17804-35-2 |
| Färg | Colorless |
| Smältpunkt | Decomposes before melting. |
| Kokpunkt | Decomposes below the boiling point. |
| Synonym | Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate |
| Industry Type | Environmental Analysis |
| Molekylvikt (g/mol) | 290.3 |
| Kvantitet | 100 mg |
| Visa mer |