Läs mer
Benomyl, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benomyl |
| CAS | 17804-35-2 |
| Molekylformel | C14 H18 N4 O3 |
| Kvantitet | 100 g |
| Synonym | 2-Benzimidazolecarbamic acid, 1-(butylcarbamoyl)-, methyl ester (8CI), Carbamic acid, [1-[(butylamino)carbonyl]-1H-benzimidazol-2-yl]-, methyl ester (9CI), Agrocit, BBC, BC 6597, Benfungin, Benfungin WP, Benlate, Benlate 50, Benlate 50W, Benlate 50WP, Benlate WP, Benomil, Benomyl, Benomyl-Imex, Du Pont 1991, Fundazol, Fundazol 50WP, Fungicide D-1991, Fungochrom, Kribenomyl, MBC, Marvel, Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, Methyl 1-(butylcarbamoyl)-2-benzimidazolylcarbamate, NS 02, NS 02 (fungicide), NSC 263489, Safomyl, Tersan 1991, Uzgen, Zetamil |
| InChI formel | InChI=1S/C14H18N4O3/c1-3-4-9-15-13(19)18-11-8-6-5-7-10(11)16-12(18)17-14(20)21-2/h5-8H,3-4,9H2,1-2H3,(H,15,19)(H,16,17,20) |
| LEDER | CCCCNC(=O)n1c(NC(=O)OC)nc2ccccc12 |
| IUPAC-namn | methyl N-[1-(butylcarbamoyl)benzimidazol-2-yl]carbamate |
| Molekylvikt (g/mol) | 290.32 |
| Formel vikt | 290.1379 |