Läs mer
Benoxacor, TRC
Produktkod. 30138179
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138179 50 mg
30134921 250 mg
17941235 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138179 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB16530550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98730-04-2
Molekylformel C11 H11 Cl2 N O2
Molekylvikt (g/mol) 260.12
Synonym 2H-1,4-Benzoxazine, 4-(dichloroacetyl)-3,4-dihydro-3-methyl- (9CI), 2,2-Dichloro-1-(2,3-dihydro-3-methyl-4H-1,4-benzoxazin-4-yl)ethanone, 4-Dichloroacetyl-3,4-dihydro-3-methyl-2H-1,4-benzoxazine, Benoxacor, CGA 154281
IUPAC-namn 2,2-dichloro-1-(3-methyl-2,3-dihydro-1,4-benzoxazin-4-yl)ethanone
LEDER CC1COc2ccccc2N1C(=O)C(Cl)Cl
Specifikationer
Kemiskt namn eller material Benoxacor
Analyt- eller komponentnamn Benoxacor
Frakt skick Room Temperature
InChI formel InChI=1S/C11H11Cl2NO2/c1-7-6-16-9-5-3-2-4-8(9)14(7)11(15)10(12)13/h2-5,7,10H,6H2,1H3
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 50 mg
Formel vikt 259.0167