Läs mer
Benoxinate Hydrochloride, TRC
Produktkod. 30348599
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30348599 5 g
1 options
Produktkod. 30348599 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1614005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Oxybuprocaine Hydrochloride
CAS 5987-82-6
Molekylformel C17 H28 N2 O3 . Cl H
Kvantitet 5 g
Synonym Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-amino-3-butoxy-, 2-(diethylamino)ethyl ester, monohydrochloride (8CI,9CI), 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate hydrochloride, Benoxil, Benoxinate hydrochloride, Boxinate, Cebesine, Conjucaine, Conjuncain, Dorsacaine, Dorsacaine hydrochloride, Lacrimin, Novesin, Novesine, Oxybuprocaine hydrochloride
InChI formel InChI=1S/C17H28N2O3.ClH/c1-4-7-11-21-16-13-14(8-9-15(16)18)17(20)22-12-10-19(5-2)6-3;/h8-9,13H,4-7,10-12,18H2,1-3H3;1H
LEDER Cl.CCCCOc1cc(ccc1N)C(=O)OCCN(CC)CC
IUPAC-namn 2-(diethylamino)ethyl 4-amino-3-butoxybenzoate;hydrochloride
Molekylvikt (g/mol) 344.88
Formel vikt 344.1867