Läs mer
Bensulide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Bensulide |
| CAS | 741-58-2 |
| Molekylformel | C14 H24 N O4 P S3 |
| Kvantitet | 250 mg |
| Synonym | Phosphorodithioic acid, O,O-diisopropyl ester, S-ester with N-(2-mercaptoethyl)benzenesulfonamide (7CI,8CI), Bensulide, Betasan, Disan, Disan (pesticide), N-[2-(O,O-Diisopropyldithiophosphoryl)ethyl]benzenesulfonamide, O,O-Diisopropyl 2-(benzenesulfonamido)ethyl dithiophosphate, O,O-Diisopropyl S-(2-benzenesulfonylaminoethyl) phosphorodithioate, Prefar, R 4461, S-β-(Benzenesulfonamido)ethyl O,O-diisopropyl dithiophosphate, SAP, SAP (herbicide) |
| InChI formel | InChI=1S/C14H24NO4PS3/c1-12(2)18-20(21,19-13(3)4)22-11-10-15-23(16,17)14-8-6-5-7-9-14/h5-9,12-13,15H,10-11H2,1-4H3 |
| LEDER | CC(C)OP(=S)(OC(C)C)SCCNS(=O)(=O)c1ccccc1 |
| IUPAC-namn | N-[2-di(propan-2-yloxy)phosphinothioylsulfanylethyl]benzenesulfonamide |
| Molekylvikt (g/mol) | 397.51 |
| Formel vikt | 397.0605 |