Learn More
Lenz Laborglasinstrumente™ Bent Adapter
Allows for tubing connections. Lenz Laborglasinstrumente™ Bent Adapters feature a bent glass olive at 90°.
Brand: Lenz Laborglasinstrumente™ 05095001
Additional Details : Weight : 0.00900kg
Description
- Constructed from DURAN™ tubing
- With NS socket
- Bore: 2.5 mm
Specifications
Bent Cone Adapter | |
Duran Glass | |
NS 14/23 |