Läs mer
Benz[e]acephenanthrylene, TRC
Produktkod. 30344203
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30344203 100 mg
1 options
Produktkod. 30344203 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB209865100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzo[b]fluoranthene
CAS 205-99-2
Molekylformel C20 H12
Kvantitet 100 mg
Synonym Benz[e]acephenanthrylene, 2,3-Benzfluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, 3,4-Benzofluoranthene, BbF, Benzo[b]fluoranthene, Benzo[e]fluoranthene, NSC 89265
InChI formel InChI=1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H
LEDER c1ccc2c(c1)c3cccc4c5ccccc5cc2c34
IUPAC-namn Benzo[b]fluoranthene
Molekylvikt (g/mol) 252.31
Formel vikt 252.0939