missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde dimethyl acetal, 99%, Thermo Scientific Chemicals

Catalog Number p-7081160
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
1125-88-8
C9H12O2
152.193
MFCD00008491
HEVMDQBCAHEHDY-UHFFFAOYSA-N
benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
62375
dimethoxymethylbenzene
COC(C1=CC=CC=C1)OC
This item is not returnable. View return policy
1125-88-8
C9H12O2
152.193
MFCD00008491
HEVMDQBCAHEHDY-UHFFFAOYSA-N
benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
62375
dimethoxymethylbenzene
COC(C1=CC=CC=C1)OC

Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY
Density 1.014
Boiling Point 196°C
Flash Point 69°C (156°F)
Odor Characteristic
Refractive Index 1.4935
Quantity 100 g
Beilstein 2044501
Solubility Information Insoluble in water. Soluble in alcohol.
Formula Weight 152.2
Percent Purity 99%
Chemical Name or Material Benzaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'