missing translation for 'onlineSavingsMsg'
Läs mer

Benzaldehyde, Sigma-Aldrich™

Produktkod. 32237680
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
32237680 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 32237680 Leverantör Sigma Aldrich Leverantörsnummer 8017569025
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Benzaldehyde
CAS 100-52-7
Smältpunkt -26°C
Densitet 1.05 g/cm³ at 20°C
Kokpunkt 179°C (1013 hPa)
Flampunkt 63°C (closed cup)
Kvantitet 25 L
Synonym Benzaldehyde, Bitter almond oil, Benzaldehyde
Molekylvikt (g/mol) 106.12
Kvalitet Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.