Läs mer
Benzamil, Tocris Bioscience™
NCX inhibitor, Deg/ENaC channel blocker; amiloride (Cat. No. 0890) derivative
Brand: Tocris Bioscience 3380/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Na+/Ca2+ exhanger (NCX) inhibitor (IC50 ∽ 100 nM); TRPP3 channel blocker (IC50 = 1.1 μM for inhibition of Ca2+-activated TRPP3 channel activity). Also non-selective Deg/ENaC family blocker; reduces mechanosensitivity of colonic afferents. More potent derivative of amiloride (Cat. No. 0890).Specifikationer
| Benzamil | |
| C13H14ClN7O·HCl | |
| N-(Benzylamidino)-3, 5-diamino-6-chloropyrazinecarboxamide hydrochloride | |
| >98% |
| 161804-20-2 | |
| 50mg | |
| 356.21 |